E.Z.N.A.® Total RNA Kit I
Lieferant: OMEGA BIO-TEK
R6834-02
R6834-01
R6834-00
R6834-03
R6834-02EA
983
EUR
R6834-02
R6834-01
R6834-00
OMEGR6834-03
E.Z.N.A.® Total RNA Kit I
Nukleinsäure-Reagenzien
Reinigungskits und Reagenzien für Nukleinsäure
Isolate total RNA from cells or soft tissues using spin column.
- Rapid – 20 minutes or less
- Safe – No phenol/chloroform extractions
- Versatile – Spin and vacuum formats available
- High quality – Purified RNA suitable for a variety of downstream applications
The E.Z.N.A.® Total RNA Kit I provides a straightforward and rapid method for the isolation of up to 100µg of total RNA from cultured eukaryotic cells and soft tissues. Multiple samples of up to 1×7 eukaryotic cells or 30mg of tissue can be processed in parallel in fewer than 20 minutes. Purified RNA can be used in many downstream applications such as RT-PCR, Northern blotting, nuclease protection assay, and in vitro translation.