Renin Substrate 1, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme: Arg-Glu(EDANS)-Ile-His-Pro-Phe-His-Leu-Val-Ile-His-Thr-Lys(DABCYL)-Arg
R8276-1MG
R8276-1MGEA 662 EUR
R8276-1MG
Renin Substrate 1, Sigma-Aldrich®
Renin Substrate 1
Substrate for human renin, an aspartic protease which plays an important role in blood-pressure regulation.