Rhodamine B isothiocyanate–Dextran
Lieferant: SIGMA ALDRICH MICROSCOPY
M-Clarity™ Qualitätsstufe = MQ200
Rhodamine B is a thiol-reactive and photostable fluorescent dye. Rhodamine B isothiocyanate is an aminoxanthene that possesses carboxylic acid and isothiocyanate substituents on the pendant phenyl ring.
Rhodamine B isothiocyanate (NE)-labeled dextran is often used as a fluorescent probe for studies of endocytotic activity and to detect the migrated of marked cells. It is also suitable for perfusion studies in animals. Rhodamine B isothiocyanate–dextran has been used for in vivo zebrafish blood-brain barrier (BBB) dye diffusion assay. Dextran labeled with rhodamine B isothiocyanate for possible use in perfusion studies in animals.
Rhodamine B stains the tissue in red-violet color and emits a bright red color when fluoresced. It is highly preferred for marking insects.
Spezifikation Testergebnisse
Appearance (Colour) Light pink to red to very dark purple |
Conforms |
Appearance (Form) | Powder |
Solubility (Colour) | Pink |
Solubility (Turbidity) 10 mg/mL, H2O |
Clear |
Degree of Substitution Mole RITC per Mole Glucose |
0.002 - 0.010 |
Average Molecular Weight of Dextran |
9000 - 11000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...