MCDh 1
Lieferant: RAL
MCDh 1 - BLEND OF NEUTRAL STAINS Ethanol based
- MCDh (Micro Chromatic Detection for Hematology)
- for RAL Stainer Kit (Ref 360200)
- Methanol-free
Role
- Fixes the smear preparation for the staining of water
- Prepares for the staining of water-soluble elements (ieBasophil granulations)
- Influences the overall intensity of the staining
MCDh is a panoptic May-Grünwald Giemsa (MGG) type staining method. Methanol free, it is made of a sequence of four specific reagents use in blood and bone marrow cell differentiation and counting. This new generation of stains is in line with the standardisation trend in diagnostic techniques. Unlike more traditional stains, all MCDh reagents are fast and ready to use. As no dilution is needed, consistency and standardisation are guaranteed.
Components - Name (ref) Packing
MCDH 1 (Ref 313590) 1L - 2,5L
MCDH 2 (Ref 313570) 1L - 2,5L
MCDH 3 (Ref 313560) 1L - 2,5L
MCDH 4 (Ref 313600) 1L - 2,5L
![]() |