Kryoboxen, PC, mit 2D Barcode und Rastereinsätzen, RATIOLAB
Lieferant: RATIOLAB
5150053
RATI5150053EA 0 EUR
RATI5150053
Kryoboxen, PC, mit 2D Barcode und Rastereinsätzen, RATIOLAB
Boxen Kryoboxen
This cryoboxes made of high quality colored polycarbonate (PC) which is used to store the Cryo-Tubes 2D.
  • For use with liquid nitrogen
  • Numerically coded
  • Stackable for safe transport
  • Autoclavable
  • Temperature-resistant from –190 °C to +121 °C
This has automatic identification of each box and its positioning through laser engraved Code 128 on the side of the lower part of the rack and 2D datamatrix code, on the bottom of the box.
Bestellinformation: Scanner and software on request.
Lieferumfang: Each pack of boxes is supplied with a vial picker.