Cover mat for deep 96-well plates
Lieferant: RATIOLAB
8101101
RATI8101101EA 278.52 EUR
RATI8101101
Cover mat for deep 96-well plates
Versiegeln Mikrotiterplatten-Dichtungen
Closures for 96-Well Deep-Well-Plates Low-Profile, 1,2 ml. Made of thermo-plastic elastomer (TPE).
Autoclavable at 121 °C, 20 minutes.