Agar

Lieferant: G Bioscienes

Synonyme: Agar Agar

RC-004 RC-003
RC-004EA 764 EUR
RC-004 RC-003
Agar
Agar
Schmelzpunkt: 90
MDL: MFCD00081288
CAS-Nummer: 9002-18-0
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR