Guanidiniumchlorid

Lieferant: G Bioscienes
Achtung

BC88 BC85 RC-057
BC88EA 435 EUR
BC88 BC85 RC-057
Guanidiniumchlorid
Guanidiniumchlorid

A selection of strong chemicals to aid in the denaturation of proteins. Denaturants, or chaotropes, disrupt water interactions resulting in the solubilization of hydrophobic proteins and peptides. The chaotropes also act on all proteins to unfold them and alter their three-dimensional structure.


Available as a powder or 8M solution

Formel: NH₂C(=NH)NH₂·HCl
Schmelzpunkt: 185 °C
Dichte: 1,354 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013026
CAS-Nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR