L(+)-Lysin Monohydrochlorid

Lieferant: G Bioscienes

RC-066 RC-067
RC-066EA 81.1 EUR
RC-066 RC-067
L(+)-Lysin Monohydrochlorid
L(+)-Lysin Monohydrochlorid
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Lagertemperatur: Gefrierschrank
MDL: MFCD00064564
CAS-Nummer: 657-27-2
EINECS: 213-091-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR