N,N,N',N'-Tetramethylethylendiamin (TEMED) ≥99,9% für die Elektrophorese
Lieferant: G Bioscienes
|
Gefahr
|
RC-101
RC-102
RC-101EA
194
EUR
RC-101
RC-102
N,N,N',N'-Tetramethylethylendiamin (TEMED) ≥99,9% für die Elektrophorese
N,N,N',N'-Tetramethylethylendiamin (TEMED)
TEMED (N,N,N’,N’-Tetramethylethylenediamine) is a 99,9% pure polymerisation reagent used when making polyacrylamide gels used in gel electrophoresis.
Formel:
C₆H₁₆N₂ Schmelzpunkt: –64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008335 CAS-Nummer: 110-18-9 UN: 2372 ADR: 3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...