Tricin Puffersubstanz

Lieferant: G Bioscienes

Synonyme: N-(Tris(hydroxymethyl)methyl)glycin

RC-104 RC-103
RC-104EA 735 EUR
RC-104 RC-103
Tricin Puffersubstanz
Tricin

This tricine is a buffer component used for the separation of low molecular weight peptides.

Formel: (HOCH₂)₃CNHCH₂CO₂H
Schmelzpunkt: 187 °C
Dichte: 1,523 g/cm³ (19.85 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004277
CAS-Nummer: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR