Dry buffer packs, JAW™
Lieferant: G Bioscienes
786-288 786-289 RC-147 RC-148
786-288EA 399 EUR
786-288 786-289 RC-147 RC-148
Dry buffer packs, JAW™
Puffer Elektrophorese-Puffer Waschpuffer
Premixed buffer powders that simply require adding water to generate a ready-to-use buffer.