Carbonate Borax Free Buffer Solution
Lieferant: REAGECON
1090BF 110025BF
REGE1090BFEA 53.6 EUR
REGE1090BF REGE110025BF
Carbonate Borax Free Buffer Solution
Puffer pH-Puffer
This range of Reagecon pH Buffers are sodium tetraborate (Borax) free. They are not subject to classification by EU regulatory authorities as toxic to reproduction under CLP category 1B. Similar in approach to the existing range of Reagecon Mercury Free Buffers, these Buffer Solutions have been developed to enhance user choice and allay safety concerns, without any impact on fitness for purpose or product performance.
  • Non toxic and borax free
  • Reagecon pH buffers are pre-programmed into the instruments of most major manufacturers
  • Laboratory chemical
For increased confidence in their test measurements, analysts should regularly measure the pH of a control standard. If an acceptable value is obtained from the control standard measurement then the analysts can have improved confidence that their test measurements will be correct. Reagecon’s extensive range of pH Buffers means that there will be a Reagecon pH Buffer which can be used as a control buffer for all pH applications.
Manufactured to exacting specifications with an extended shelf life and stability, even after opening the bottle thereby eliminating the requirement to open a fresh bottle of solution every time it is used.
Standards: Reagecon’s Borax free pH buffer solutions are tested by our ISO 17025 accredited test method and NIST traceable.
Verpackung: HDPE bottles.