Ammoniumcer(IV)nitrat 0.1 mol/l (0.1 N) in wässriger Lösung Reag. Ph. Eur.
Lieferant: REAGECON
|
Achtung
|
Synonyme:
Diammoniumhexanitratocerat, di-Ammonium-hexanitratocerat (IV), Cer(IV)ammoniumnitrat
3000100
REGE3000100EA
433
EUR
REGE3000100
Ammoniumcer(IV)nitrat 0.1 mol/l (0.1 N) in wässriger Lösung Reag. Ph. Eur.
Ammoniumcer(IV)nitrat
Formel:
(NH₄)₂Ce(NO₃)₆ Molecular Weight: 548,22 g/mol |
MDL:
MFCD00151121 CAS-Nummer: 16774-21-3 EINECS: 240-827-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...