Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) Reag. Ph. Eur. Chapter 4 (4.2.2)
Lieferant: REAGECON
|
Gefahr
|
Reagecon's Tetrabutylammonium Hydroxide 0.1M is a fully factorised, high purity, stable product, developed and tested for titrations. This Analytical Volumetric Solution, also called titrant, standard titrant or standard solution, is tested to a specification of ± 0.2%. It is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
The product may be used for manual titrations, or more commonly automatic titrations, to determine the end points by detection of any of several properties as detailed in several EP monographs.
Formel:
C₁₆H₃₇NO Lagertemperatur: Kühlschrank |
MDL:
MFCD00009425 CAS-Nummer: 2052-49-5 EINECS: 218-147-6 UN: 3286 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...