Zinksulfat 0.1 mol/l (0.1 N) aqueous solution Reag. Ph. Eur. 3008600
Lieferant: REAGECON
|
|
3008600
REGE3008600EA
108
EUR
REGE3008600
Zinksulfat 0.1 mol/l (0.1 N) aqueous solution Reag. Ph. Eur. 3008600
Zinksulfat
Formel:
O₄SZn Molecular Weight: 161.45 g/mol Schmelzpunkt: 100 Dichte: 1.45 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011302 CAS-Nummer: 7733-02-0 EINECS: 231-793-3 UN: 3082 ADR: 9 III Merck Index: 12,10293 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...