Arsenic standard solution, for limit tests
Lieferant: REAGECON
5000500C
REGE5000500CEA 135 EUR
REGE5000500C
Arsenic standard solution, for limit tests
Standards Kationischer Standards Arsen-Standards
Concentrate to make arsenic standard solution (10 ppm as As) as outlined in Chapter 4 (4.1.2) of Ph. Eur.