Ammoniumthiocyanat 0,1 M according to (ChP)

Lieferant: REAGECON

Synonyme: Ammoniumsulfocyanid, Ammoniumrhodanid

CPAT2010F
REGECPAT2010FEA 68.6 EUR
REGECPAT2010F
Ammoniumthiocyanat 0,1 M according to (ChP)
Ammoniumthiocyanat

Reagecon's Ammonium Thiocyanate 0,1 M according to Chinese Pharmacopoeia (ChP) is a fully factorised, high purity, stable product, developed and tested for titrations. This is an Analytical Volumetric Solution, also called titrant, standard titrant or standard solution. It is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete. The product may be used for manual titrations, or more commonly automatic titrations also called potentiometric titration, to determine the end points by detection of any of several properties as detailed in several ChP monographs.

Formel: NH₄SCN
Molecular Weight: 76.12 g/mol
Schmelzpunkt: 150
MDL: MFCD00011428
CAS-Nummer: 1762-95-4
Merck Index: 13,00564

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR