Turbidity standards, Very low range 0 to 1 NTU
Lieferant: REAGECON
CRS-0.1-100
CRS-0.1-500
CRS-0.2-100
CRS-0.2-500
CRS-0.5-500
CRS-1-100
CRS-1-500
CRSR-0.1-100
CRSR-0.1-500
CRSR-0.2-100
CRSR-0.2-500
CRSR-0.4-100
CRSR-0.4-500
CRSR-0.5-100
CRSR-0.5-500
CRSR-1-100
CRSR-1-500
CRS-0.0-100
CRS-0.0-500
REGECRS-0.1-100EA
128
EUR
REGECRS-0.1-100
REGECRS-0.1-500
REGECRS-0.2-100
REGECRS-0.2-500
REGECRS-0.5-500
REGECRS-1-100
REGECRS-1-500
REGECRSR-0.1-100
REGECRSR-0.1-500
701-0048
REGECRSR-0.2-500
REGECRSR-0.4-100
REGECRSR-0.4-500
REGECRSR-0.5-100
REGECRSR-0.5-500
REGECRSR-1-100
REGECRSR-1-500
REGECRS-0.0-100
REGECRS-0.0-500
Turbidity standards, Very low range 0 to 1 NTU
Standards
Trübungsstandards
Turbidity standards for ratio and non-ratio instruments composed of suspended polymer microspheres.
- Non toxic and non carcinogenic microspheres
- Highly accurate with well defined spherical shape
- Traceable to NIST
- US EPA approved
- Ready to use- particles stay in suspension