Leitfähigkeitsstandard 133 µS/cm (20°C; KCl)
Lieferant: REAGECON
EP133 EP1330 EP266
REGEEP133EA 285 EUR
REGEEP133 REGEEP1330 REGEEP266
Leitfähigkeitsstandard 133 µS/cm (20°C; KCl)
Standards Leitfähigkeits-Standards
A complete range of certified control standards with values ranging from 133 to 26,6 µS.
Standards: Standards are manufactured and tested with the compliance with the European Pharmacopeia.
Lieferumfang: Standards are supplied with certificate of analysis and safety data sheet.