Leitfähigkeitsstandard 133 µS/cm (20°C; KCl)
Lieferant: REAGECON
EP133
EP1330
EP266
REGEEP133EA
285
EUR
REGEEP133
REGEEP1330
REGEEP266
Leitfähigkeitsstandard 133 µS/cm (20°C; KCl)
Standards
Leitfähigkeits-Standards
A complete range of certified control standards with values ranging from 133 to 26,6 µS.
Standards: Standards are manufactured and tested with the compliance with the European Pharmacopeia.
Lieferumfang: Standards are supplied with certificate of analysis and safety data sheet.