Gasoline Calibration Standard (3 Compound Mix) in Petroleum Ether
Lieferant: REAGECON
Gasoline calibration standard (three compound mix) in petroleum ether is a ready to use standard used in fuel analysis as either a calibrant or analytical control solution. This standard is used in testing for a range of Aromatic and oxygenate compounds in gasoline in accordance with the ASTM D5845, using infrared (IR) instrumentation. The product can also be used to validate appropriate test methods or qualify an instrument in a regulated industry. The product is QC tested on a high performance GC-MS. This product is prepared gravimetrically on a weight/volume basis to a specification of ±2.5%
- Multi compound mix analyte
- Mixed concentrations
- Weight: 0,57 kg
- Packing group II with class three
Standards: Reagecon holds ISO 17025 accreditation (A2LA Ref: 6739.02) for calibration of laboratory balances.