Carbon dioxide standard solutions
Lieferant: REAGECON
SC023C SC022C SC026C SC024C
REGESC023CEA 354 EUR
REGESC023C REGESC022C REGESC026C REGESC024C
Carbon dioxide standard solutions
Standards Umweltanalyse-Standards
Standards for M965D carbon dioxide analyser, for soft drinks and brewery applications.