Natriumthiosulfat 0.1 mol/l (0.1 N) aqueous solution Volumetric

Lieferant: REAGECON

Synonyme: di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit

T20105 T20101
REGET20105EA 76.4 EUR
REGET20105 REGET20101
Natriumthiosulfat 0.1 mol/l (0.1 N) aqueous solution Volumetric
Natriumthiosulfat
Formel: Na₂S₂O₃
Molecular Weight: 158.11 g/mol
Siedepunkt: 100
Schmelzpunkt: 48
Lagertemperatur: Raumtemperatur
MDL: MFCD00003499
CAS-Nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR