Carbon standards
Lieferant: REAGECON
TOIC10 TOIC100 TOIC1M TOC50 TIC1M TOC1M TIC500 TOC500 TOC2M TOC5 TIC5 TOC100 TOC10 TOC20 TOC15 TOC250
REGETOIC10EA 369 EUR
REGETOIC10 REGETOIC100 701-0049 REGETOC50 REGETIC1M REGETOC1M REGETIC500 REGETOC500 REGETOC2M REGETOC5 REGETIC5 REGETOC100 REGETOC10 REGETOC20 REGETOC15 REGETOC250
Carbon standards
Standards Kationischer Standards Kohlenstoff-Standards
These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.
These standards are independent, traceable and certified.
Standards: Accredited to IEC/ISO 17025 and ISO 17034.