Carbon standards
Lieferant: REAGECON
TOIC10
TOIC100
TOIC1M
TOC50
TIC1M
TOC1M
TIC500
TOC500
TOC2M
TOC5
TIC5
TOC100
TOC10
TOC20
TOC15
TOC250
REGETOIC10EA
369
EUR
REGETOIC10
REGETOIC100
701-0049
REGETOC50
REGETIC1M
REGETOC1M
REGETIC500
REGETOC500
REGETOC2M
REGETOC5
REGETIC5
REGETOC100
REGETOC10
REGETOC20
REGETOC15
REGETOC250
Carbon standards
Standards
Kationischer Standards
Kohlenstoff-Standards
These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.
These standards are independent, traceable and certified.
Standards: Accredited to IEC/ISO 17025 and ISO 17034.