Blei(II)acetat

Lieferant: REAGECON

Synonyme: Essigsäure Blei(II)salz, Blei(II)acetat

USP3105
REGEUSP3105EA 201 EUR
REGEUSP3105
Blei(II)acetat
Blei(II)acetat

Standards: ISO 17025 Accredited Test Method (A2LA Ref: 6739.03).

Formel: (H₃CCOO)₂Pb
Molecular Weight: 325.29 g/mol
Schmelzpunkt: 280
MDL: MFCD00012452
CAS-Nummer: 301-04-2
UN: 3082
ADR: 9 III
Merck Index: 12,05411

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR