Methylorange USP

Lieferant: REAGECON

USP3405
REGEUSP3405EA 56.8 EUR
REGEUSP3405
Methylorange USP
Methylorange
Formel: C₁₄H₁₄N₃NaO₃S
MDL: MFCD00007502
CAS-Nummer: 547-58-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR