Calibration Cuvettes
Lieferant: REICHERT
13103410
REIC13103410EA 909 EUR
REIC13103410
Calibration Cuvettes
Küvetten Spezialküvetten
Re-usable glass calibration cuvette with special option coatings assayed at a mid-range Total Serum Bilirubin value. Used to perform an automatic calibration on the Reichert UNISTAT® Bilirubinometer.
Approximate Assay Value 20 mg/dL, 342 µmol/L