Secondary columns for GCxGC (fused silica), Rtx-200
Lieferant: Restek
15111
REST15111EA 133 EUR
REST15111
Secondary columns for GCxGC (fused silica), Rtx-200
Chromatographiesäulen
Convenient 2 m length is ideal for use as a secondary column in GCxGC analyses.
  • General purpose column
  • midpolarity phase; Crossbond trifluoropropylmethyl polysiloxane
  • Temperature range: –20 to 340 °C
The Rtx-200 column offers exceptional thermal stability, low bleed, and superior inertness—even for active compounds such as phenols, and with sensitive detectors such as ECDs, NPDs, and MSDs.