Tubing reamer
Lieferant: Restek
20134
REST20134EA 55 EUR
REST20134
Tubing reamer
Werkzeugsätze
The tubing reamer removes burrs from stainless steel tubing.
  • For 3,1 to 6,35 mm tubing
  • Nonslip safety design