Tubing cutter
Lieferant: Restek
20192
REST20192EA 146 EUR
REST20192
Tubing cutter
Schneidewerkzeuge
Designed to cut hard or soft copper, aluminium, brass, stainless steel, Monel® alloy, or titanium.
  • Produces square, smooth cuts in ¹/₁₆" tubing
  • Eliminates tubing distortion