Graphite O-ring for Shimadzu 2010 and 2014 GCs
Lieferant: Restek
20243
REST20243EA 80 EUR
REST20243
Graphite O-ring for Shimadzu 2010 and 2014 GCs
Versiegeln Dichtringe
Graphite O-ring are for use with numerous Shimadzu gas chromatography systems.
  • Available in packs of 5
  • Durable graphite construction
  • Max operating temperature of 450 °C
For use with Shimadzu 2010 and 2014 GC systems.