Inlet liner packing tool
Lieferant: Restek
20339
REST20339EA 120 EUR
REST20339
Inlet liner packing tool
Werkzeug zum Herausziehen und Einführen
The inlet liner packing tool can be used with all different liners.
  • Accurate to a specific measured depth
  • Position wool reproducibly every time