Open end wrench set
Lieferant: Restek
20387
REST20387EA 155 EUR
REST20387
Open end wrench set
Schraubenschlüssel
High-quality 1/4×5/16", 3/8"×7/16", 7/16"×1/2", and 1/2"×9/16" wrenches for tightening a wide variety of chromatography fittings.