Polyimide, Harz

Lieferant: Restek

20445
REST20445EA 50 EUR
REST20445
Polyimide, Harz
Polyimide

Polyimide resin securely connects a press-tight connector to a fused silica column.


  • Max temperature: 350 °C

Formel: C₃₅H₂₈N₂O₇
MDL: MFCD01868102
CAS-Nummer: 62929-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR