Backpressure regulator kit
Lieferant: Restek
20634
REST20634EA 0 EUR
REST20634
Backpressure regulator kit
Druckminderer
This backpressure regulator kit increases the versatility of Agilent 5890 GC by replacing the existing 0,06 bar (207 kPa) backpressure regulator and gauge with a 4,14 bar regulator.
  • Enables to use longer 60 and 105 m columns as well as shorter 10 m columns
Lieferumfang: Kit is supplied with complete instructions.