Capillary GC inlet systems have backpressure regulators to maintain a constant upstream pressure and rapidly respond to catastrophic leaks. The operating range 0 to 4,1 bar is sufficient to operate a 105 m, 0,25 mm ID column at its optimum flow rate.