Indicating hydrocarbon traps for air compressors
Lieferant: Restek
20637 20636
REST20637EA 0 EUR
REST20637 REST20636
Indicating hydrocarbon traps for air compressors
Filter Gasfilter
These indicating hydrocarbon traps for air compressors remove oil vapours and mist as it passes compressed air from an oil-filled air compressor. Indicator changes from pale pink to deep pink.
Dimensions: 152×44 mm