GC capillary column connectors kit
Lieferant: Restek
20645
REST20645EA 107 EUR
REST20645
GC capillary column connectors kit
Fittings und Verbinder
This split or splitless inlet adaptor fitting is suitable for capillary columns with internal diameter 0,53 mm. Makes column installation easy due to the nut protruding farther from the insulated injection port chamber.
  • Accepts standard two-hole capillary ferrules and a standard 1/8 inch nut
  • Column insertion depth is the same as the original equipment
  • Use recessed gooseneck liners with this adaptor
Bestellinformation: This kit is supplied with adaptor fitting, capillary nut, stainless steel inlet seal, washer, and one 0,8 mm ID two-hole ferrule.