Single stage gas regulators
Lieferant: Restek
20665 20666 20667
REST20665EA 1626 EUR
REST20665 REST20666 REST20667
Single stage gas regulators
Druckminderer
These stainless steel regulators are the standard for ultra high purity and corrosion resistant pressure regulation. They are more easily purged of atmospheric components compared to brass gas regulators.
  • Use when there is secondary pressure regulation downstream
  • Identical gas purity protection as with dual-stage gas regulators
  • Especially useful in atmospheres of dry corrosive gases such as hydrogen
  • Outlet assembly: 316 L stainless steel diaphragm valve and ¹/₄ tube fitting