Injector wrench for Shimadzu
Lieferant: Restek
21159
REST21159EA 49 EUR
REST21159
Injector wrench for Shimadzu
Schraubenschlüssel
This stainless steel inlet wrench is ideal for Shimadzu 17A, 2010, and 2014 GCs.
  • Designed specifically for removing Shimadzu injection ports