Cylinder valve wrench
Lieferant: Restek
21321
REST21321EA 169 EUR
REST21321
Cylinder valve wrench
Schraubenschlüssel
This specially designed wrench enables easy opening of cylinder valves that are fitted with a hand wheel. It is also suitable for removing difficult cylinder caps.