Universal cylinder wrench
Lieferant: Restek
21322
REST21322EA 88.5 EUR
REST21322
Universal cylinder wrench
Schraubenschlüssel
This versatile wrench is used for tightening gauges and gas regulator CGA fittings to cylinder outlets and pipe thread connections.