Hydrogen flashback arrestor
Lieferant: Restek
21334
REST21334EA 598 EUR
REST21334
Hydrogen flashback arrestor
Druckminderer
This hydrogen flashback arrestor is a special gas safety device, it cuts to stop the flame or reverse flow of gas back up into the equipment or supply line and it prevents the user and equipment from damage or explosions.
  • Valve cuts off gas supply in the event of a flashback
  • Can be reset and reused after flashback has occurred
  • Valve spring is made of 316 stainless steel
Maximum pressure: Hydrogen, 3,5 bar, temperature sensor: polyethylene
Standards: Meets safety requirements of BS6158, EN730, ISO 5175, AS 4603 and are UL Listed 23Y5.