Piston seal insertion tool, HPLC pumps
Lieferant: Restek
21356
REST21356EA 85 EUR
REST21356
Piston seal insertion tool, HPLC pumps
Werkzeug zum Herausziehen und Einführen
Piston seal insertion tool is an alternative for paper clip and screw to remove the worn seals from the HPLC pump.
  • Simplify pump maintenance
  • Removes old piston seal and installs new seal
  • It cannot mar the surrounding metal surface of the pump housing