Mini regulator
Lieferant: Restek
22032
REST22032EA 611 EUR
REST22032
Mini regulator
Druckminderer
This mini regulator is suitable for natural gas and refinery gas standards.
Inlet pressure range: 0 to 20,68 bar
Outlet pressure range: 0 to 1,03 bar
Lieferumfang: Supplied with outlet pressure gauge (0 - 1,03 bar), brass CGA 170 nut and nipple, and compression outlet (3,175 mm).