Injector wrench for Agilent
Lieferant: Restek
22065
REST22065EA 55 EUR
REST22065
Injector wrench for Agilent
Schraubenschlüssel
This stainless steel injector wrench is uses to remove the septum nut and weldments during GC maintenance.
  • Smaller end to remove the septum nut
  • Larger end to tighten the split/splitless weldment nut