Dual Vespel® ring cross-disk inlet seals
Lieferant: Restek
22084
REST22084EA 447 EUR
REST22084
Dual Vespel® ring cross-disk inlet seals
Versiegeln Mechanische Dichtungen
These inlet seals are washerless and leak-tight and ideal for high-flow split applications >500 ml/min.