Single-Stage Ultra-High Purity Chrome-Plated Brass Gas Regulators with International Fittings
Lieferant: Restek
22138
REST22138EA 756 EUR
REST22138
Single-Stage Ultra-High Purity Chrome-Plated Brass Gas Regulators with International Fittings
Druckminderer
UHP brass regulators are the best choice when using ultra-high-purity carrier gas for sensitive GC applications using MS, PID, or ECD detection methods.
  • Use when there is secondary pressure regulation downstream
  • Identical gas purity protection as with dual-stage gas regulators
  • Oxidation-resistant
  • Chrome-plated
All regulators are rated to 206 bar maximum inlet pressure. Chrome plated brass materials also includes BS 341 #3 (He, Ar, Air, N2); single-stage type.