Make life easier (MLE) capillary tool kit for Shimadzu GCs
Lieferant: Restek
22182
REST22182EA 581 EUR
REST22182
Make life easier (MLE) capillary tool kit for Shimadzu GCs
Werkzeugsätze
MLE tool kits conveniently provide the tools that make it easier to install and maintain capillary columns.
Lieferumfang: Kit includes ¹/₈", ³/₁₆", ¹/₄" nylon brushes, ¹/₄", ³/₈", ³/₁₆" stainless steel wire tube brushes, stainless steel surface brush, 6 stainless steel jet reamers (0,25 to 0,65 mm OD), ¹/₄×⁵/₁₆" open end wrench, ³/₈×⁷/₁₆" open end wrench, rubber-tipped slide-lock tweezers, scoring wafers, inlet liner removal tool, septum puller, mini wool puller/inserter tool, 4-inch tapered needle file, swivel head flashlight, mini hand drill set, 15 cm compact steel ruler, pocket magnifier, high temperature string (1 m), pipe cleaner (12-inch), cotton tip swabs (pack of 25), capillary installation gauge, injector wrench, 6×7 mm open end wrench, 8×10 mm open end wrench and 16×17 mm open end wrench.